Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578105 Supplier Invitrogen™ Supplier No. PA578105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: MCF-7 Predicted Reactivity: Bovine (82%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The superfamily of high molecular weight serine proteinase inhibitors. A critical portion of the molecule, the reactive center loop connects beta sheets A and C. Protease inhibitor-8 (PI8; SERPINB8) is a member of the ov-serpin subfamily, which, relative to the archetypal serpin PI1 (MIM 107400), is characterized by a high degree of homology to chicken ovalbumin, lack of N- and C-terminal extensions, absence of a signal peptide, and a serine rather than an asparagine residue at the penultimate position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene SERPINB8
Gene Accession No. P50452
Gene Alias CAP2; CAP-2; Cytoplasmic antiproteinase 2; NK10; ovalbumin; peptidase inhibitor 8; PI8; PI-8; protease inhibitor 8 (ovalbumin type); serine (or cysteine) peptdiase inhibitor, clade B, member 8; serine (or cysteine) peptidase inhibitor, clade B, member 8; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 8; serine (or cysteine) proteinase inhibitor, clade B, member 8; serine protease inhibitor 8; serpin B8; serpin family B member 8; serpin peptidase inhibitor, clade B (ovalbumin), member 8; Serpinb8; Spi8
Gene Symbols SERPINB8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SerpinB8. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.