Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110355
Description
Immunogen sequence: EQFDRKYTRG MLFKTNEEKK TVQMMFKEAK FKMGYADEVH TQVLELPYVE EELSMVILLP DDNTDLAVKE.
The superfamily of high molecular weight serine proteinase inhibitors. A critical portion of the molecule, the reactive center loop connects beta sheets A and C. Protease inhibitor-8 (PI8; SERPINB8) is a member of the ov-serpin subfamily, which, relative to the archetypal serpin PI1 (MIM 107400), is characterized by a high degree of homology to chicken ovalbumin, lack of N- and C-terminal extensions, absence of a signal peptide, and a serine rather than an asparagine residue at the penultimate position.
Specifications
| SERPINB8 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB8 | |
| CAP2; CAP-2; Cytoplasmic antiproteinase 2; NK10; ovalbumin; peptidase inhibitor 8; PI8; PI-8; protease inhibitor 8 (ovalbumin type); serine (or cysteine) peptdiase inhibitor, clade B, member 8; serine (or cysteine) peptidase inhibitor, clade B, member 8; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 8; serine (or cysteine) proteinase inhibitor, clade B, member 8; serine protease inhibitor 8; serpin B8; serpin family B member 8; serpin peptidase inhibitor, clade B (ovalbumin), member 8; Serpinb8; Spi8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20725, 288937, 5271 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O08800, P50452 | |
| SERPINB8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 173-242 of human SERPINB8 (NP_001027018.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction