Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583122 Supplier Invitrogen™ Supplier No. PA583122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YFKGKWNEPF DETYTREMPF KINQEEQRPV QMMYQEATFK LAHVGEVRAQ LLELPYARKE LSLLV Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 65%.

PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SERPINB9
Gene Accession No. P50453
Gene Alias BB283241; CAP3; CAP-3; Cytoplasmic antiproteinase 3; ovalbumin; peptidase inhibitor 9; PI9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) peptidase inhibitor, clade B, member 9; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; serine (or cysteine) proteinase inhibitor, clade B, member 9; serine protease inhibitor 6; serpin B9; serpin family B member 9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; SERPINB9; Spi6; testicular tissue protein Li 180
Gene Symbols SERPINB9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERPINB9. Recombinant protein control fragment (Product #RP-94947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.