Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB9 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | SERPINB9 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119075
|
Invitrogen™
PA5119075 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121615
|
Invitrogen™
PA5121615 |
20 μL |
Each for $209.50
|
|
|||||
Description
SERPINB9 Polyclonal Antibody for Western Blot, IHC (P)
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).Specifications
SERPINB9 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
SERPINB9 | |
BB283241; CAP3; CAP-3; Cytoplasmic antiproteinase 3; ovalbumin; peptidase inhibitor 9; PI9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) peptidase inhibitor, clade B, member 9; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; serine (or cysteine) proteinase inhibitor, clade B, member 9; serine protease inhibitor 6; serpin B9; serpin family B member 9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; SERPINB9; Spi6; testicular tissue protein Li 180 | |
SERPINB9 | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P50453 | |
5272 | |
Recombinant protein Cytoplasmic Antiproteinase 3. The antigen corresponds to amino acid range 1-376 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title