Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582862 Supplier Invitrogen™ Supplier No. PA582862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSEAINELTV LVLVNTIYFK GLWKSKFSPE NTRKELFYKA DGESCSASMM YQEGKFRYRR VAEGTQVLEL PFKGDDITMV LILPKPEKSL AKVEKELTPE VLQEWLDELE EMMLVVHMPR FRIEDGFSLK EQLQDMGLV Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 88%.

The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Serpinc1
Gene Accession No. P01008
Gene Alias AI114908; Antithrombin; Antithrombin 3; anti-thrombin 3; antithrombin III; antithrombin-III; AT3; At-3; AT3D; ATIII; hypothetical protein LOC540261; MGC22579; PRO0309; serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1; serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1; serpin C1; serpin family C member 1; serpin peptidase inhibitor clade C member 1; serpin peptidase inhibitor, clade C (antithrombin), member 1; serpin peptidase inhibitor, clade C, member 1; Serpinc1; THPH7
Gene Symbols Serpinc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERPINC1. Recombinant protein control fragment (Product #RP-99383).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.