Learn More
Invitrogen™ SERPINC1 Polyclonal Antibody

Description
PA1-43054 detects SERPINC1 in human samples. PA1-43054 has successfully been used in Western blot and ELISA applications. PA1-43054 was derived from human antithrombin III.
Specifications
Specifications
| Antigen | SERPINC1 |
| Applications | ELISA, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Concentration | 14.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | Serpinc1 |
| Gene Accession No. | P01008 |
| Gene Alias | AI114908; Antithrombin; Antithrombin 3; anti-thrombin 3; antithrombin III; antithrombin-III; AT3; At-3; AT3D; ATIII; hypothetical protein LOC540261; MGC22579; PRO0309; serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1; serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1; serpin C1; serpin family C member 1; serpin peptidase inhibitor clade C member 1; serpin peptidase inhibitor, clade C (antithrombin), member 1; serpin peptidase inhibitor, clade C, member 1; Serpinc1; THPH7 |
| Gene Symbols | Serpinc1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.