Learn More
Invitrogen™ SERPINE2 Monoclonal Antibody (OTI2C9)

Description
SERPINE2 Monoclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | SERPINE2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide, pH 7.3 |
| Gene | SERPINE2 |
| Gene Accession No. | P07093 |
| Gene Alias | B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.