Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINE2 Monoclonal Antibody (OTI6F3), TrueMAB™, OriGene
SDP

Catalog No. 502540156
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0156 30 μL
50-167-5344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0156 Supplier OriGene Supplier No. TA504718S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINE2 Monoclonal antibody specifically detects SERPINE2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SERPINE2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6F3
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FXYD3
Gene Accession No. Q14802
Gene Alias B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4
Host Species Mouse
Immunogen Full length human recombinant protein of human FXYD3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5349
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.