Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587074
Description
Purity is > 95% by SDS-PAGE.
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
SERPINF2 | |
Polyclonal | |
Unconjugated | |
SERPINF2 | |
A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18816, 287527, 5345 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P08697, Q61247 | |
SERPINF2 | |
Recombinant fusion protein of human SERPINF2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction