Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588240 Supplier Invitrogen™ Supplier No. PA588240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEMVQLGAKG KAQQQIRQTL KQQETSAGEE FFVLKSFFSA ISEKKQEFTF NLANALYLQE GFTVKEQYLH GNKEFFQSAI KLVDFQDAKA CAEMISTWVE RKTDGKIKDM FSGEEFGPLT RLVLVNAIYF KGDWKQKFRK EDTQLINFTK KNGSTVKIPM MKALLRTKYG YFSESSLNYQ VLELSYKGDE FSLIIILPAE GMDIEEVEKL ITAQQILKWL SEMQEEEVEI SLPRFKVEQK VDFKDVLYSL NITEIFSGGC DLSGITDSSE VYVSQVTQKV FFEINEDGSE AATSTGIHIP VIMSLAQSQF IANHPFLFIM KHNPTESILF MGRVTNPDTQ EIKGRDLDSL; Positive Samples: Mouse pancreas, Rat pancreas.

The protein encoded by this gene is a member of the serine protease inhibitor superfamily made up of proteins which play central roles in the regulation of a wide variety of physiological processes, including coagulation, fibrinolysis, development, malignancy and inflammation. The gene product may have a role in a growth-control, possibly growth-suppressing pathway and, when impaired, may be involved in pancreatic carcinogenesis. The protein is a member of the plasminogen activator inhibitor-1 family, a subset of the serpin superfamily whose members act as tissue-specific tPA inhibitors. Two alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINI2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SERPINI2
Gene Accession No. Q9JK88
Gene Alias MEPI; Myoepithelium-derived serine protease inhibitor; neuroserpin; PANCPIN; Pancreas-specific protein TSA2004; pancreas-specific serpin; peptidase inhibitor 14; PI14; PI-14; protease inhibitor 14; serine (or cysteine) peptidase inhibitor, clade I, member 2; serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 2; serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2; serine protease inhibitor 14; serpin family I member 2; serpin I2; serpin peptidase inhibitor, clade I (pancpin), member 2; serpin peptidase inhibitor, clade I, member 2; SERPIN12; SERPINI2; Spi14; TSA2004
Gene Symbols SERPINI2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171149, 67931
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.