Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serratia marcescens Monoclonal Antibody (B/N4N)
GREENER_CHOICE

Catalog No. PIMA546567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546567 Supplier Invitrogen™ Supplier No. MA546567
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serratia marcescens Monoclonal Antibody for ICC/IF, ELISA

Serratia marcescens (Sma) is a Gram-negative bacterium that is now recognized as an important opportunistic pathogen. It causes a range of nosocomial infections, such as pneumonia, urinary tract infections, bacteremia, meningitis and endocarditis, particularly in immunocompromised patients [PMID: 35315751].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serratia marcescens
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone B/N4N
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 10mM sodium azide
Gene Alias S. marcescens
Host Species Mouse
Immunogen Native Serratia marcescens (ATCC 13880).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.