Learn More
Sertaconazole nitrate, Thermo Scientific™
Specifications
Specifications
| Chemical Name or Material | Sertaconazole nitrate |
| CAS | 99592-39-9 |
| Quantity | 1 g |
| Melting Point | 156°C to 165°C |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C20 H15 Cl3 N2 O S . H N O3 |
| Physical Form | Powder |
| Color | White to Beige |
| Purity | ≥97.5% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.