Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERTAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113449
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113449 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113449 Supplier Invitrogen™ Supplier No. PA5113449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERTAD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SEI-1, also known as SERTAD1 (SERTA domain containing 1) or TRIP-Br1 (Transcriptional regulator interacting with the PHD-bromodomain 1), is a transcriptional regulator that integrates signals provided by transcription factors. Acting at E2F-responsive promoters, SEI-1 interacts with the PHD- and bromodomains of proteins such as TIF1 and DP-1, thereby transmitting their signals to the promoter and stimulating transcriptional activity. SEI-1 exists as a multiprotein complex with E2F-1 and DP-1 and is expressed at different levels throughout the cell cycle, allowing it to regulate cell cycle progression via promoter control during the G1 and S phases. Additionally, SEI-1 can render the activity of the cyclin D-Cdk4 complex, an important catalyst of the cell cycle, resistant to the inhibitory effects of p16. Overexpression of SEI-1 is implicated in the development of squamous cell carcinomas of the head and neck.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERTAD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SERTAD1
Gene Accession No. Q9UHV2
Gene Alias 1110032C13Rik; AV000455; AW456625; CDK4-binding protein p34SEI; CDK4-binding protein p34SEI1; p34SEI-1; PHD zinc finger- and bromodomain-interacting protein 1; SEI1; SEI-1; SERTA domain containing 1; SERTA domain-containing protein 1; SERTAD1; Transcriptional regulator interacting with the PHD-bromodomain 1; TRIPBR1; TRIP-Br1
Gene Symbols SERTAD1
Host Species Rabbit
Immunogen Recombinant Human SERTA domain-containing protein 1 protein (1-236AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29950
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.