Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERTAD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543700
Description
Peptide sequence: LQRQTIFNIS LMKLYNHRPL TEPSLQKTVL INNMLRRIQE ELKQEGSLRP Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.
Acts at E2F-responsive promoters as coregulator to integrate signals provided by PHD- and/or bromodomain-containing transcription factors. May act as coactivator as well as corepressor of E2F1-TFDP1 and E2F4-TFDP1 complexes on E2F consensus binding sites, which would activate or inhibit E2F-target genes expression. Modulates fat storage by down-regulating the expression of key genes involved in adipocyte lipolysis, thermogenesis and oxidative metabolism. [UniProt].
Specifications
SERTAD2 | |
Polyclonal | |
Unconjugated | |
SERTAD2 | |
AB041541; AU021878; Kiaa0127; MGC126688; MGC126690; mKIAA0127; MNCb-1504; PHD zinc finger- and bromodomain-interacting protein 2; Sei2; Sei-2; SERTA domain containing 2; SERTA domain-containing protein 2; SERTAD2; Transcriptional regulator interacting with the PHD-bromodomain 2; transcriptional regulator interacting with the PHS-bromodomain 2; TRIPBR2; TRIP-Br2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9792 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14140 | |
SERTAD2 | |
Synthetic peptide directed towards the N-terminal of human SERTAD2 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction