Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERTAD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987240
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7240 Supplier Bioss Supplier No. BS6398R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acts at E2F-responsive promoters to integrate signals provided by PHD- and/or bromodomain-containing transcription factors.

Specifications

Antigen SERTAD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SERTAD2
Gene Accession No. Q14140
Gene Alias AB041541; AU021878; KIAA0127; LOC100340129; MGC126688; MGC126690; mKIAA0127; MNCb-1504; PHD zinc finger- and bromodomain-interacting protein 2; Sei2; SEI-2; SERTA domain containing 2; SERTA domain-containing protein 2; SERTAD2; Transcriptional regulator interacting with the PHD-bromodomain 2; transcriptional regulator interacting with the PHS-bromodomain 2; TRIPBR2; TRIP-Br2
Gene Symbols SERTAD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SERTAD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9792
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.