Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERTM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567107
Description
Immunogen sequence: SGSVENGTFLE LFPTSLSTSV DPSSGHLSN Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
SERTM1 is a protein coding gene.
Specifications
SERTM1 | |
Polyclonal | |
Unconjugated | |
SERTM1 | |
C13orf36; serine rich and transmembrane domain containing 1; serine-rich and transmembrane domain containing 1; serine-rich and transmembrane domain-containing 1; serine-rich and transmembrane domain-containing protein 1; SERTM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
400120 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2A2V5 | |
SERTM1 | |
Recombinant Human SERTM1. Recombinant protein control fragment (Product #RP-107751). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction