Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid A (SAA1) Monoclonal Antibody (OTI7A4), TrueMAB™, OriGene
SDP

Catalog No. 502543504
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3504 Supplier OriGene Supplier No. CF813993
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serum Amyloid A (SAA1) Monoclonal antibody specifically detects Serum Amyloid A (SAA1) in Human samples. It is validated for ELISA

Specifications

Antigen Serum Amyloid A (SAA1)
Applications ELISA
Classification Monoclonal
Clone OTI7A4
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MPO
Gene Accession No. P05164
Gene Alias amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4
Host Species Mouse
Immunogen Full length recombinant protein of human MPO (NP_000241) produced in Baculovirus-Insect cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.