Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A1/A2 Antibody (SR1117), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Serum Amyloid A1/A2 |
---|---|
Clone | SR1117 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Serum Amyloid A1/A2 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
6288 | |
IgG | |
Affinity purified |
SR1117 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4 | |
A synthesized peptide derived from human Serum Amyloid A1/A2 (Uniprot #: P0DJI8) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title