Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A1/A2 Mouse anti-Human, Clone: SAA/326, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30776620UG
Description
Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Serum Amyloid A1/A2 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Mouse | |
Protein A or G purified | |
RUO | |
6288 | |
Store at 4°C. | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
SAA/326 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction