Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A1 Antibody (CL9175), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Serum Amyloid A1 |
---|---|
Clone | CL9175 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Serum Amyloid A1 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
This antibody was developed against a Synthetic Peptide of P0DJI8. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL9175 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience | |
PBS, pH 7.2, 40% glycerol | |
6288 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title