Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A1 Antibody (Reu86.5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311246
Description
Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.Specifications
Serum Amyloid A1 | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay | |
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
The antibody is raised against human SAA and Helix Pomatia Haemocyanine. | |
50 μg | |
Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience | |
6288 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
Reu86.5 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction