Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A1 Antibody (SAA1cc), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073071B200UL
Description
Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Human, Canine samples. It is validated for Western Blot, ELISA.Specifications
Serum Amyloid A1 | |
Monoclonal | |
Biotin | |
PBS | |
SAA1 | |
human recombinant SAA, conjugated with carrier protein. | |
Alzheimers Research, Cancer, Neuroscience | |
Human recombinant SAA | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA | |
SAA1 | |
Western Blot, ELISA | |
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
Mouse | |
200 μL | |
6288 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction