Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serum Amyloid A4 Antibody (014), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289368MFV450
Description
Serum Amyloid A4 Monoclonal antibody specifically detects Serum Amyloid A4 in Human samples. It is validated for ELISASpecifications
Serum Amyloid A4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
014 | |
Sandwich ELISA Detection | |
C-SAACSAAConstitutively expressed serum amyloid A protein, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serum Amyloid A4 (Uniprot#: P35542; Glu19-Tyr130). | |
0.1 mL | |
Immunology | |
6291 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction