Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serum Amyloid P Monoclonal Antibody (273912)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA523975

Catalog No. PIMA523975


Only null left
Add to Cart

Description

Description

In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2 and 25% cross-reactivity with recombinant mouse (rm) Pentraxin 2 is observed. In direct ELISAs, no cross-reactivity with rhPentraxin 2 or rmPentraxin 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Amyloid P
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P23680
APCS
Mouse myeloma cell line NS0-derived recombinant rat Pentraxin2/SAP Gln21-Ser228 (predicted).
100 μg
Primary
Rat
Antibody
IgG2b
Western Blot
273912
Unconjugated
APCS
9.5S alpha-1-glycoprotein; amyloid P component, serum; APCS; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; PTX2; SAMP; Sap; Serum amyloid P-component; Serum amyloid P-component(1-203)
Mouse
Protein A/G
RUO
29339
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.