Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102543 Supplier Thermo Scientific Supplier No. PA5102543
Add to Cart
Add to Cart

Serum Amyloid P Polyclonal Antibody

Serum Amyloid P Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Serum Amyloid P
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APCS
Gene Accession No. P02743, P12246, P23680
Gene Alias 9.5S alpha-1-glycoprotein; amyloid P component, serum; APCS; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; PTX2; SAMP; Sap; Serum amyloid P-component; Serum amyloid P-component(1-203)
Gene Symbols APCS
Host Species Rabbit
Immunogen A synthesized peptide derived from human APCS(Accession P02743), corresponding to amino acid residues L138-I188.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20219, 29339, 325
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.