Learn More
Invitrogen™ Serum Amyloid P Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596028
Description
Positive Samples: 293T Immunogen sequence: MNKPLLWISV LTSLLEAFAH TDLSGKVFVF PRESVTDHVN LITPLEKPLQ NFTLCFRAYS DLSRAYSLFS YNTQGRDNEL LVYKERVGEY SLYIGRHKVT SKVIEKFPAP VHICVSWESS SGIAEFWING TPLVKKGLRQ GYFVEAQPKI VLGQEQDSYG GKFDRSQSFV GEIGDLYMWD SVLPPENILS AYQGTPLPAN ILDWQALNYE IRGYVIIKPL VWV.
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
Specifications
Serum Amyloid P | |
Polyclonal | |
Unconjugated | |
APCS | |
9.5S alpha-1-glycoprotein; amyloid P component, serum; APCS; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; PTX2; SAMP; Sap; Serum amyloid P-component; Serum amyloid P-component(1-203) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20219, 29339, 325 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P02743, P12246, P23680 | |
APCS | |
A synthetic peptide corresponding to a sequence within amino acids 1-223 of human APCS (NP_0016301). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.