Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One VACUETTE™ Z Serum Blood Collection Tubes

Catalog No. p-9393004
Click to view available options
Capacity (Metric):
9 mL
Closure Color:
Red with Black Ring
This item is not returnable. View return policy
This item is not returnable. View return policy

VACUETTE™ Z Serum Blood Collection Tubes are used in determinations for clinical chemistry, immunology and serology. Available in Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator. The inner wall of the tube is coated with silica particles which activate clotting when the tubes are turned.

  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for clinical chemistry, immunology and serology testing
  • Specially coated with microscopic silica particles, which activate the coagulation process
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Available in: Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator
  • CE marked
TRUSTED_SUSTAINABILITY

Specifications

Additive Z Serum Clot Activator
Material PET
Sterility Sterile
Closure Type Non-ridged Pull Cap
Capacity (Metric) 9 mL
For Use With (Equipment) VACUETTE™ Blood Collection Needles
Closure Color Red with Black Ring
Tube Size 16 x 100 mm
For Use With (Application) Blood Collection
Length (Metric) 100 mm
Diameter (Metric) Outer 16 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.