Learn More
Labconco™ Service Fixture Kits
SDP
Catalog No. 09039047 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Cold Water (CW) GN Fixture Kit
Cold Water (CW) Service Fixture Kit
Gas (GAS) Service Fixture Kit
HW/CW Mixing GN Fixture Kit
Nitrogen (NIT) Service Fixture Kit
Oxygen (OXY) Service Fixture Kit
Steam (STM) Service Fixture Kit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
09-039-047 Nitrogen (NIT) Service Fixture Kit
09-039-046 Steam (STM) Service Fixture Kit
09-039-048 Oxygen (OXY) Service Fixture Kit
09-039-050 HW/CW Mixing GN Fixture Kit
09-039-039 Cold Water (CW) Service Fixture Kit
09-039-042 Gas (GAS) Service Fixture Kit
09-039-049 Cold Water (CW) GN Fixture Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-039-047 Supplier Labconco™ Supplier No. 9809300
Add to Cart
Add to Cart
Kit includes copper tubing, brass valve, knob handle and outlet.
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and tubing, color-coded fixture knob.
Specifications
For Use With (Equipment) Protector XLE Laboratory Hood
Weight (English) 4 lb.
Weight (Metric) 1.8 Kg
Includes 0.25 in. dia. Copper Tubing, Brass Valve and Green Knob, Connector
Product Type Nitrogen (NIT) Service Fixture Kit
For Use With (Application) Nitrogen service
WARNING: Reproductive Harm - www.P65Warnings.ca.gov