Learn More
BrandTech™ VACUUBRAND™ Service Kits for Oil-Free Diaphragm Vacuum Pumps
Original VACUUBRAND replacement parts
$231.84 - $1414.56
Specifications
Type Service Kit
View More Specs
Products 7
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
13-688-970
View Documents
BrandTech™ VACUUBRAND™
20696863
MZ2D NT VARIO
Each for $403.20
Only null left
Add to Cart
13-889-108
View Documents
BrandTech™ VACUUBRAND™
20696878
ME2C NT Diaphragm Pump
Each for $316.96
Only null left
Add to Cart
13-688-969
View Documents
BrandTech™ VACUUBRAND™
20696862
ME 8 NT
Each for $692.72
Only null left
Add to Cart
13-688-968
View Documents
BrandTech™ VACUUBRAND™
20696861
MD 4 NT/VARIO, MV 2 NT/VARIO
Each for $716.24
Only null left
Add to Cart
13-688-967
View Documents
BrandTech™ VACUUBRAND™
20696860
MZ 2 NT, ME 4 NT + VARIO
Each for $443.52
Only null left
Add to Cart
13-889-107
View Documents
BrandTech™ VACUUBRAND™
20696874
ME1, ME1C Diaphragm Pump
Each for $231.84
Only null left
Add to Cart
13-889-106
View Documents
BrandTech™ VACUUBRAND™
20696828
MD1C, PC3001 Diaphragm Pump
Each for $1,414.56
Only null left
Add to Cart
Description
Description
  • Includes appropriate diaphragms, valves, and sealing rings, if appropriate
  • Does not include spanner wrench
Specifications
Specifications
Service Kit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.