Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ One/OneC On-site Installation & Recertification

Catalog No. 701058111
Click to view available options
Type:
Annual OQ Recertification
Security Suite Install + IQ/OQ
For Use With (Equipment):
NanoDrop One/OneC

Rely on our trained experts to perform on-site installation or recertification of your Thermo Scientific™ NanoDrop™ One/OneC. Services include on-site installation of NanoDrop One/OneC PC Software and 21 CFR Part 11 compliance software, basic user orientation, instrument installation qualification and operational qualification. All engineer labor and travel charges for installation within continental US.

Rely on our trained experts to perform on-site installation or recertification of your Thermo Scientific™ NanoDrop™ One/OneC. Services include on-site installation of NanoDrop One/OneC PC Software and 21 CFR Part 11 compliance software, basic user orientation, instrument installation qualification and operational qualification. All engineer labor and travel charges for installation within continental US. Additional charges may apply for outside of continental US or remote Canadian regions. Separate purchase of 21 CFR Part 11 compliance software for NanoDrop One/OneC Spectrophotometers and NanoDrop One/OneC IQ/OQ Kit required.

NanoDrop One/OneC Validated Installation

Features:

  • On-site installation of instrument
  • On-site installation of NanoDrop One PC Control and Security Suite software
  • Basic user orientation
  • Instrument installation Qualification and Operational Qualification
  • All engineer labor and travel charges for installation within continental US. Additional charges may apply for outside of continental US or remote Canadian regions.

Requires:

  • Separate purchase of Security Suite for NanoDrop One/OneC
  • Separately purchased NanoDrop One IQ/OQ Kit
  • Separately purchased Qualification Filter Kit for NanoDrop 2000C and
  • OneC (for NanoDrop OneC instruments only)

Exclusions:

  • Consumables such as cuvettes, pipettes, flash drives, printer paper

NanoDrop Digital Signature SW OQ

Features:

  • Execution of NanoDrop One/OneC Security Suite OQ

Requires:

  • Separate purchase of Security Suite for NanoDrop One/OneC
  • Requires the purchase of 701-058108 NanoDrop One/OneC Validated Installation

 

NanoDrop One/OneC OQ Recertification

  • Annual Operational Qualification
  • All engineer labor and travel charges for OQ within continental US. Additional charges may apply for outside of continental US or remote Canadian regions.

Requires:

  • Separate purchase of Security Suite for NanoDrop One/OneC
  • Separately purchased NanoDrop One IQ/OQ Kit or NanoDrop One Requalification Kit
  • Separately purchased Qualification Filter Kit for NanoDrop 2000C and OneC (for NanoDrop OneC instruments only)

Exclusions:

  • Consumables such as cuvettes, pipettes, flash drives, printer paper

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance 21 CFR Part 11
Display Digital
Type Annual OQ Recertification
Wavelength Range 190 to 850 nm
For Use With (Application) Spectrophotometry
For Use With (Equipment) NanoDrop One/OneC
Includes All engineer labor and travel charges for OQ within continental US. Additional charges may apply for outside of continental US or remote Canadian regions. Requires separate purchase of NanoDrop One IQ/OQ Kit (and cuvette filter kit) or NanoDrop One Requalification Kit (PN 840-282800)
Sample Volume (Metric) 1 to 2 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.