Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Servus™ Neoprene III with Neo-Grip Soles

Catalog No. p-3660119
Click to view available options
Size:
6
7
8
9
11
13
Height (Metric):
30.48 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

100% waterproof seamless construction provides comfort, flexibility and excellent traction

  • Unique dipping process makes boots soft and pliable for less leg fatigue
  • Neo-Grip™ outsoles provide excellent traction on wet surfaces to reduce injuries from slips and falls
  • Removable Breathe-O-Prene™ insoles provide shock absorption and moisture-wicking comfort
  • Tough, comfortable fabric lining
  • Available in copper tan Mid Pac and Hi Pac
  • Steel toe

Mid Pac

  • Height is 30.48cm (12 in.)
  • US men's sizes 6-13
  • Hi Pac
    • Height is 40.6cm (16 in. )
    • US men's sizes 6, 8-9, 11, 13

    Compliance

    Meets electrical hazard requirements of ASTM F2413-05. Steel toe also meets ASTM F2413-05. Neoprene styles meet ASTM F2413-11 EH

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI Z41 PT 99
    Color Tan
    Height (English) 12 in.
    Height (Metric) 30.48 cm
    Size 6
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.