Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Servus™ Neoprene III with Neo-Grip Soles
Click to view available options
Size:
6
7
8
9
11
13
Height (Metric):
30.48 cm
Description
- Unique dipping process makes boots soft and pliable for less leg fatigue
- Neo-Grip™ outsoles provide excellent traction on wet surfaces to reduce injuries from slips and falls
- Removable Breathe-O-Prene™ insoles provide shock absorption and moisture-wicking comfort
- Tough, comfortable fabric lining
- Available in copper tan Mid Pac and Hi Pac
- Steel toe
Mid Pac
- Height is 30.48cm (12 in.)
- US men's sizes 6-13
- Height is 40.6cm (16 in. )
- US men's sizes 6, 8-9, 11, 13
Compliance
Meets electrical hazard requirements of ASTM F2413-05. Steel toe also meets ASTM F2413-05. Neoprene styles meet ASTM F2413-11 EH

Specifications
Specifications
Certifications/Compliance | ANSI Z41 PT 99 |
Color | Tan |
Height (English) | 12 in. |
Height (Metric) | 30.48 cm |
Size | 7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction