Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seryl tRNA synthetase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157539
Description
Seryl tRNA synthetase Polyclonal specifically detects Seryl tRNA synthetase in Human samples. It is validated for Western Blot.Specifications
Seryl tRNA synthetase | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 6.1.1.11, FLJ36399, serine-tRNA ligase, Serine--tRNA ligase, SERRS, SERSserine tRNA ligase 1, cytoplasmic, Seryl-tRNA Ser/Sec synthetase, seryl-tRNA synthetase, seryl-tRNA synthetase, cytoplasmic, Seryl-tRNA(Ser/Sec) synthetase | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Pig: 92%; Rat: 92%; Xenopus: 85%; Zebrafish: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q5T5C8 | |
SARS | |
Synthetic peptides corresponding to SARS (seryl-tRNA synthetase) The peptide sequence was selected from the middle region of SARS. Peptide sequence SQFDEELYKVIGKGSEKSDDNSYDEKYLIATSEQPIAALHRDEWLRPEDL. | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
6301 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction