Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seryl-tRNA synthetase Polyclonal Antibody
Seryl-tRNA synthetase Polyclonal Antibody
Supplier: Thermo Scientific PA529761
Description
Seryl-tRNA synthetase Polyclonal Antibody for Western Blot, IHC (P), IP

Specifications
Seryl-tRNA synthetase | |
Polyclonal | |
Unconjugated | |
SARS1 | |
EC 6.1.1.11; SARS; Sars1; serine tRNA ligase 1, cytoplasmic; serine--tRNA ligase, cytoplasmic; SerRS; SERS; seryl-aminoacyl-tRNA synthetase; seryl-aminoacyl-tRNA synthetase 1; seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; seryl-tRNA(Ser/Sec) synthetase; Strs | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20226, 266975, 6301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P26638, P49591, Q6P799 | |
SARS1 | |
Recombinant fragment corresponding to a region within amino acids 28 and 372 of Human Seryl-tRNA synthetase. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction