Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Seryl-tRNA synthetase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566705
Description
Immunogen sequence: GSRGYIPIYTP FFMRKEVMQE VAQLSQFDEE LYKVIGKGSE KSDDNSYDEK YLIATSEQPI AALHRDEWLR PEDLPIKYAG LSTCFRQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.
Seryl-tRNA synthetase (SerRS) catalyzes the attachment of serine to tRNA(Ser) in a two-step reaction: serine is first activated by ATP to form Ser-AMP and then transferred to the acceptor end of tRNA(Ser). Is also probably able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).
Specifications
Seryl-tRNA synthetase | |
Polyclonal | |
Unconjugated | |
SARS1 | |
EC 6.1.1.11; SARS; Sars1; serine tRNA ligase 1, cytoplasmic; serine--tRNA ligase, cytoplasmic; SerRS; SERS; seryl-aminoacyl-tRNA synthetase; seryl-aminoacyl-tRNA synthetase 1; seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; seryl-tRNA(Ser/Sec) synthetase; Strs | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49591 | |
SARS1 | |
Recombinant Human Seryl-tRNA synthetase. Recombinant protein control fragment (Product #RP-105329). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction