Learn More
Invitrogen™ Seryl-tRNA synthetase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593183
Description
Immunogen sequence: MVLDLDLFRV DKGGDPALIR ETQEKRFKDP GLVDQLVKAD SEWRRCRFRA DNLNKLKNLC SKTIGEKMKK KEPVGDDESV PENVLSFDDL TADALANLKV SQIKKVRLLI DEAILKCDAE RIKLEAERFE NLREIGNLLH PSVPISNDED VDNKVERIWG DCTVRKKYSH VDLVVMVDGF EGEKGAVVAG SRGYFLKGVL VFLEQALIQY ALRTLGSRGY IPIYTPFFMR KEVMQEVAQL SQFDEELYKV IGKGSEKSDD NSYDEKYLIA TSEQPIAALH RDEWLRPEDL PIKYAGLSTC; Positive Samples: MCF7, HepG2, HeLa, U-251MG, 293T, Mouse kidney, Mouse brain, Rat kidney, Rat brain; Cellular Location: Cytoplasm.
Seryl-tRNA synthetase (SerRS) catalyzes the attachment of serine to tRNA(Ser) in a two-step reaction: serine is first activated by ATP to form Ser-AMP and then transferred to the acceptor end of tRNA(Ser). Is also probably able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).
Specifications
Seryl-tRNA synthetase | |
Polyclonal | |
Unconjugated | |
SARS1 | |
EC 6.1.1.11; SARS; Sars1; serine tRNA ligase 1, cytoplasmic; serine--tRNA ligase, cytoplasmic; SerRS; SERS; seryl-aminoacyl-tRNA synthetase; seryl-aminoacyl-tRNA synthetase 1; seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; seryl-tRNA(Ser/Sec) synthetase; Strs | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20226, 266975, 6301 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26638, P49591, Q6P799 | |
SARS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SARS (NP_0065042). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.