Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seryl-tRNA synthetase Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA527615

Catalog No. PIPA527615


Description

Description

PA5-27615 targets seryl-tRNA synthetase in WB applications and shows reactivity with Human samples. The PA5-27615 immunogen is recombit fragment corresponding to a region within amino acids 347 and 514 of Human seryl-tRNA synthetase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene belongs to the class II amino-acyl tRNA family. The encoded enzyme catalyzes the transfer of L-serine to tRNA (Ser) and is related to bacterial and yeast counterparts.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

seryl-tRNA synthetase
Polyclonal
Unconjugated
P49591
SARS
Recombit fragment corresponding to a region within amino acids 347 and 514 of Human seryl-tRNA synthetase
100 μL
Primary
Human
Antibody
IgG
Western Blot
0.23 mg/mL
SARS
SERRS, SERS, Seryl-tRNA Ser/Sec synthetase, serine tRNA ligase 1, cytoplasmic, serine--tRNA ligase, cytoplasmic, seryl-tRNA synthetase, cytoplasmic, seryl-tRNA(Ser/Sec) synthetase
Rabbit
Antigen affinity chromatography
RUO
6301
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.