Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesame oil

Catalog No. p-191936
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC241002500

Supplier: Thermo Scientific Chemicals 241002500

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 8008-74-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-74-0
MDL Number MFCD00132243

Specifications

Acidity 0.6mg KOH/g max.
Melting Point -5.0°C
Color Yellow
Density 0.9190g/mL
Flash Point 255°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4760
Quantity 250 mL
Specific Gravity 0.919
Merck Index 14, 8469
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, ether, petroleum ethers
Physical Form Viscous Liquid
Chemical Name or Material Sesame oil
Show More Show Less
EINECSNumber 232-370-6
RTECSNumber VU3940000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.