Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesame Oil, NF, Spectrum™ Chemical
SDP

Catalog No. 18606187 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
6 x 500 mL
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum

Catalog No. 18-606-187

Supplier: Spectrum Chemical Manufacturing Corporation SE130500MLGLCS6

Only null left
Add to Cart
Add to Cart

SE130, 8008-74-0

Spectrum™ Chemical Sesame Oil, NF is used as an excipient solvent carrier in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. All Spectrum Chemical NF are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8008-74-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 6 x 500 mL
Saponification Value 188-195
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.