Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sesame Oil, NF, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
6 x 500 mL
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Sesame Oil, NF is used as an excipient solvent carrier in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. All Spectrum Chemical NF are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Drum |
| Quantity | 200 L |
| Saponification Value | 188-195 |
| Grade | NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction