Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesamol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
533-31-3
C7H6O3
138.12
MFCD00005827
LUSZGTFNYDARNI-UHFFFAOYSA-N
sesamol,3,4-methylenedioxy phenol,5-hydroxy-1,3-benzodioxole,3,4-methylenedioxyphenol,2h-1,3-benzodioxol-5-ol,3,4-methylendioxyphenol,methylene ether of oxyhydroquinone,phenol, 3,4-methylenedioxy,5-benzodioxolol,unii-94iea0nv89
68289
CHEBI:9126
1,3-benzodioxol-5-ol
C1OC2=C(O1)C=C(C=C2)O
533-31-3
C7H6O3
138.12
MFCD00005827
LUSZGTFNYDARNI-UHFFFAOYSA-N
sesamol,3,4-methylenedioxy phenol,5-hydroxy-1,3-benzodioxole,3,4-methylenedioxyphenol,2h-1,3-benzodioxol-5-ol,3,4-methylendioxyphenol,methylene ether of oxyhydroquinone,phenol, 3,4-methylenedioxy,5-benzodioxolol,unii-94iea0nv89
68289
CHEBI:9126
1,3-benzodioxol-5-ol
C1OC2=C(O1)C=C(C=C2)O
Color White to Beige
Water 0.2% max. (K.F.)
Flash Point >110°C
Packaging Glass bottle
Quantity 25g
Melting Point 63.0°C to 65.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 19, 66
Solubility Information Solubility in water: slightly soluble.
Formula Weight 138.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Sesamol
Show More Show Less

Recommended Storage : Darkens on storage particularly when stored in the light and at elevated temperatures