Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesamol, 98%

Catalog No. p-3737362
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 533-31-3 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00005827
InChI Key LUSZGTFNYDARNI-UHFFFAOYSA-N
Synonym sesamol, 3,4-methylenedioxy phenol, 5-hydroxy-1,3-benzodioxole, 3,4-methylenedioxyphenol, 2h-1,3-benzodioxol-5-ol, 3,4-methylendioxyphenol, methylene ether of oxyhydroquinone, phenol, 3,4-methylenedioxy, 5-benzodioxolol, unii-94iea0nv89
PubChem CID 68289
ChEBI CHEBI:9126
IUPAC Name 1,3-benzodioxol-5-ol
SMILES C1OC2=C(O1)C=C(C=C2)O

Specifications

Melting Point 63.0°C to 65.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 19, 66
Solubility Information Solubility in water: slightly soluble.
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sesamol
Show More Show Less
EINECSNumber 208-561-5
Recommended Storage Darkens on storage particularly when stored in the light and at elevated temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.