Learn More
Description
Specifications
Specifications
| Antigen | SESN1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot >1:500, ELISA < 1:10000 |
| Formulation | Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide |
| Gene Alias | MGC138241, MGC142129, p53-regulated protein PA26, PA26p53 activated gene 26, SEST1p53 regulated PA26 nuclear protein, sestrin 1, sestrin-1 |
| Gene Symbols | SESN1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to unique epitope on SESN1. The SESN1 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
