Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SESN1 Antibody (SR2166), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322266100UL
Description
SESN1 Monoclonal antibody specifically detects SESN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
SESN1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR2166 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
MGC138241, MGC142129, p53-regulated protein PA26, PA26p53 activated gene 26, SEST1p53 regulated PA26 nuclear protein, sestrin 1, sestrin-1 | |
A synthesized peptide derived from human SESN1 (Uniprot #: Q9Y6P5 ) | |
100 μg | |
Cell Cycle and Replication | |
27244 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction