Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SESN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111712 Supplier Invitrogen™ Supplier No. PA5111712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASRFEIEKRE SMFVFSSDDE EVTPARAVSR HFEDTSYGYK DFSRHGMHVP TFRVQDYC Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 95%.

This gene encodes a member of the sestrin family. Sestrins are induced by the p53 tumor suppressor protein and play a role in the cellular response to DNA damage and oxidative stress. The encoded protein mediates p53 inhibition of cell growth by activating AMP-activated protein kinase, which results in the inhibition of the mammalian target of rapamycin protein. The encoded protein also plays a critical role in antioxidant defense by regenerating overoxidized peroxiredoxins, and the expression of this gene is a potential marker for exposure to radiation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SESN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sesn1
Gene Accession No. Q9Y6P5
Gene Alias 1110002G11Rik; AU044290; LOW QUALITY PROTEIN: sestrin-1; p53 activated gene 26; p53 regulated PA26 nuclear protein; p53-regulated protein PA26; PA26; Sesn1; Sest1; sestrin 1; sestrin-1; si:ch211-51a19.4; WITHDRAWN: zgc:103669; zgc:103669; zgc:91970
Gene Symbols Sesn1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SESN1. Recombinant protein control fragment (Product #RP-107941).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.