Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SESN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120259 Supplier Invitrogen™ Supplier No. PA5120259
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, U-87MG, HepG2, K-562. Immunogen sequence: VFGCGILPEG DADGSPAPQA PTPPSEQSSP PSRDPLNNSG GFESARDVEA LMERMQQLQE SLLRDEGTSQ EEMESRFELE KSESLLVTPS ADILEPSPHP DMLCFVEDPT FGYEDFTRRG AQAPPTFRAQ DYTWEDHGYS LIQRLYPEGG QLLDEKFQAA YSLTYNTIAM HSGVDTSVLR RAIWNYIHCV FGIRYDDYDY GEVNQLLERN LKVYIKTVAC YPEKTTRRMY NLFWRHFRHS EKVHVNLLLL EARMQAALLY ALRAITRYMT.

This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SESN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SESN2
Gene Accession No. P58004, P58043
Gene Alias Hi95; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; RGD1566319; Ses2; SESN2; SEST2; sestrin 2; sestrin-2
Gene Symbols SESN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 211-480 of human SESN2 (NP_113647.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230784, 83667
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.