Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SESN3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987045
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7045 Supplier Bioss Supplier No. BS6100R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May function as an intracellular leucine sensor that negatively regulates the TORC1 signaling pathway (PubMed:25263562). May also regulate the insulin-receptor signaling pathway through activation of TORC2 (By similarity). This metabolic regulator may also play a role in protection against oxidative and genotoxic stresses (By similarity).

Specifications

Antigen SESN3
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SESN3
Gene Accession No. P58005
Gene Alias 5630400E15Rik; BC003348; Sesn3; SEST3; sestrin 3; Sestrin-3
Gene Symbols SESN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SESN3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143686
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.