Learn More
Invitrogen™ SESTD1 Monoclonal Antibody (OTI7G4)

Description
SESTD1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | SESTD1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI7G4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Sestd1 |
| Gene Accession No. | Q80UK0, Q86VW0 |
| Gene Alias | 1500031J16Rik; A930010O20; EGK_04585; Huntingtin-interacting protein-like protein; protein Solo; RGD1562244; SEC14 and spectrin domain containing 1; SEC14 and spectrin domains 1; SEC14 domain and spectrin repeat-containing protein 1; Sestd1; SOLO; Solo/Trio8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.