Learn More
SESTD1 Mouse anti-Canine, Human, Mouse, Clone: OTI7G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505929
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.Specifications
| SESTD1 | |
| Monoclonal | |
| Unconjugated | |
| SESTD1 | |
| SOLO | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 228071, 478823, 91404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7G4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q80UK0, Q86VW0 | |
| Sestd1 | |
| Full length human recombit protein of human SESTD1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.