Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sestrin2 Mouse anti-Human, Porcine, Rat, Clone: 1A1A4, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 662971IG150UL
Description
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.Specifications
| Sestrin2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58004 | |
| Sesn2 | |
| Sestrin 2 Fusion Protein Ag20638 | |
| 150 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1A1A4 | |
| Unconjugated | |
| SESN2 | |
| HI95, SES2, SESN2, SEST2, sestrin 2, Sestrin2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100620966, 502988, 83667 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction